missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-1,5-benzodioxepin-7-amine, 97%, Thermo Scientific™

Product Code. 10180232
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10180232 10 g Amber Glass Bottle 10g
10784373 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10180232 Supplier Thermo Scientific Maybridge Supplier No. BTB12798EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175136-34-2
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00218806
InChI Key FVLCICVRAPEYNX-UHFFFAOYSA-N
Synonym 3,4-dihydro-2h-benzo b 1,4 dioxepin-7-amine, 2h-1,5-benzodioxepin-7-amine,3,4-dihydro, 2h-1,5-benzodioxepin-7-amine, 3,4-dihydro, 2h,3h,4h-benzo b 1,4-dioxepin-7-ylamine, maybridge1_002016, acmc-1c5sw, buttpark 12115-34, 3,4-trimethylenedioxyaniline, 7-amino-3,4-dihydro-2h-1,5-benzodioxepine, 3, 4-dihydro-7-amino-2h-benzo b 1,4 dioxepine
PubChem CID 2775654
IUPAC Name 3,4-dihydro-2H-1,5-benzodioxepin-7-amine
SMILES C1COC2=C(C=C(C=C2)N)OC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 165.19
Percent Purity 97%
Chemical Name or Material 3, 4-Dihydro-2H-1, 5-benzodioxepin-7-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.