missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-1,4-benzoxazine-2-carbonitrile, 97+%, Thermo Scientific™

Product Code. 10265834
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10265834 5 g Amber Glass Bottle 5g
10473413 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10265834 Supplier Thermo Scientific Maybridge Supplier No. CC29716DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 86267-86-9
Molecular Formula C9H8N2O
Molecular Weight (g/mol) 160.176
InChI Key YSTANLOUKDVPGJ-UHFFFAOYSA-N
Synonym 3,4-dihydro-2h-benzo b 1,4 oxazine-2-carbonitrile, 2h,3h,4h-benzo e 1,4-oxazaperhydroine-2-carbonitrile, 2h-1,4-benzoxazine-2-carbonitrile,3,4-dihydro
PubChem CID 2795504
IUPAC Name 3,4-dihydro-2H-1,4-benzoxazine-2-carbonitrile
SMILES C1C(OC2=CC=CC=C2N1)C#N

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 160.18
Percent Purity 97%
Chemical Name or Material 3, 4-Dihydro-2H-1, 4-benzoxazine-2-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.