missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2-(1H)-quinolinone, 98%

Product Code. 15467677
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15467677 1 g 1g
15477677 5 g 5g
15487677 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15467677 Supplier Thermo Scientific Alfa Aesar Supplier No. H36638.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 553-03-7 | C9H9NO | 147.18 g/mol

3,4-Dihydro-2-(1H)-quinolinone may be employed as medium supplement in the culture medium of Pseudomonas ayucida during enrichment culture experiments. It is used to prepare potent bicyclic peptide deformylase inhibitors with antibacterial effects. It is also used to synthesize substituted iminopiperidines as inhibitors of human nitric oxide synthase isoforms.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dihydro-2-(1H)-quinolinone may be employed as medium supplement in the culture medium of Pseudomonas ayucida during enrichment culture experiments. It is used to prepare potent bicyclic peptide deformylase inhibitors with antibacterial effects. It is also used to synthesize substituted iminopiperidines as inhibitors of human nitric oxide synthase isoforms.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 553-03-7
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00016722
InChI Key TZOYXRMEFDYWDQ-UHFFFAOYSA-N
Synonym 3,4-dihydro-2 1h-quinolinone, 3,4-dihydroquinolin-2 1h-one, hydrocarbostyril, 1,2,3,4-tetrahydroquinolin-2-one, 3,4-dihydrocarbostyril, 2-oxo-1,2,3,4-tetrahydroquinoline, dihydro-quinolinone, 3,4-dihydro-2-quinolinol, o-aminohydrocinnamic acid lactam, unii-2ckg6tx32f
PubChem CID 64796
SMILES O=C1CCC2=CC=CC=C2N1

Specifications

Melting Point 165°C to 167°C
Quantity 1 g
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
IUPAC Name 3,4-dihydro-1H-quinolin-2-one
Formula Weight 147.18
Percent Purity 98%
Chemical Name or Material 3,4-Dihydro-2-(1H)-quinolinone
compliance-icons
Product Identifier
  • 3,4-Dihydro-2-(1H)-quinolinone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 3,4-Dihydro-2(1H)-quinolinone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.