missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorotoluene, 98%

Product Code. 11388938
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388938 1 g 1g
11308948 5 g 5g
11398938 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11388938 Supplier Thermo Scientific Alfa Aesar Supplier No. 041789.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2927-34-6 | C7H6F2 | 128.12 g/mol

Used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2927-34-6
Molecular Formula C7H6F2
Molecular Weight (g/mol) 128.12
MDL Number MFCD00075087
InChI Key FZMPLKVGINKUJZ-UHFFFAOYSA-N
PubChem CID 2733400
IUPAC Name 1,2-difluoro-4-methylbenzene
SMILES CC1=CC=C(F)C(F)=C1

Specifications

Density 1.120 g/mL
Boiling Point 110°C to 113°C
Flash Point 25°C (77°F)
Odor Aromatic
Refractive Index 1.45
Quantity 1 g
UN Number UN1993
Synonym 3,4-difluorotoluene, benzene, 1,2-difluoro-4-methyl, 1,2-difluoro-4-methyl-benzene, 3,4-difluoro methyl benzene, 3,4 difluorotoluene, pubchem1615, 3, 4-difluorotoluene, 3,4-difluoro toluene
Solubility Information Insoluble in water.
Formula Weight 128.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3,4-Difluorotoluene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.