missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorophenylacetic acid, 98%, Thermo Scientific™

Product Code. 10551734 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10551734 5 g Glass bottle 5g
10207280 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10551734 Supplier Thermo Scientific Acros Supplier No. 311640050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 658-93-5
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.131
MDL Number MFCD00010002
InChI Key YCAKYFIYUHHCKW-UHFFFAOYSA-N
Synonym 3,4-difluorophenylacetic acid, 2-3,4-difluorophenyl acetic acid, 3,4-difluorophenyl acetic acid, benzeneacetic acid, 3,4-difluoro, 3,4-difluorobenzeneacetic acid, 3,4-difluorophenylaceticacid, pubchem4154, acmc-209nst, ksc355a2l, rarechem al bo 0250
PubChem CID 69571
IUPAC Name 2-(3,4-difluorophenyl)acetic acid
SMILES C1=CC(=C(C=C1CC(=O)O)F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 47°C to 51°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula F2F6H3CH2COOH
Quantity 5 g
Formula Weight 172.13
Percent Purity ≥97.5%
Physical Form Crystalline Needles
Chemical Name or Material 3, 4-Difluorophenylacetic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.