missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorophenyl isocyanate, Thermo Scientific™

Product Code. 10065433 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass Bottle in Plastic Container
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10065433 10 g Glass Bottle in Plastic Container 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10065433 Supplier Thermo Scientific Acros Supplier No. 369520100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42601-04-7
Molecular Formula C7H3F2NO
Molecular Weight (g/mol) 155.104
MDL Number MFCD00065711
InChI Key YBWJVFMNWMLZEN-UHFFFAOYSA-N
Synonym 3,4-difluorophenyl isocyanate, 3,4-difluorophenylisocyanate, isocyanic acid 3,4-difluorophenyl ester, 3,4-difluoro phenyl isocyanate, benzene,1,2-difluoro-4-isocyanato, 1,2-difluoro-4-isocyanato-benzene, 3,4-difluorobenzenisocyanate, acmc-1apvi, 3,4-difluoro-phenylisocyanate, 3,4-difluoro-phenyl isocyanate
PubChem CID 2734708
IUPAC Name 1,2-difluoro-4-isocyanatobenzene
SMILES C1=CC(=C(C=C1N=C=O)F)F

Specifications

Color Yellow
Density 1.3260g/mL
Infrared Spectrum Authentic
Packaging Glass Bottle in Plastic Container
Linear Formula F2C6H3NCO
Refractive Index 1.4970 to 1.4990 (20°C, 589nm)
Quantity 10 g
Formula Weight 155.1
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 3, 4-Difluorophenyl isocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.