missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluoronitrobenzene, 99%

Product Code. 10219213 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10219213 100 mL Glass bottle 100mL
10226820 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10219213 Supplier Thermo Scientific Acros Supplier No. 241931000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Reglering: Begränsningar kan gälla för köp av denna produkt och vi kan behöva begära ytterligare dokumentation från dig.

CAS: 369-34-6 | C6H3F2NO2 | 159.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 369-34-6
Molecular Formula C6H3F2NO2
Molecular Weight (g/mol) 159.09
MDL Number MFCD00007198
InChI Key RUBQQRMAWLSCCJ-UHFFFAOYSA-N
Synonym 3,4-difluoronitrobenzene, benzene, 1,2-difluoro-4-nitro, 1-nitro-3,4-difluorobenzene, 1,2-difluoro-4-nitro-benzene, 3,4-difluoro nitrobenzene, pubchem2290, 3,4-difluornitrobenzene, 3.4-difluoronitrobenzene, 3, 4-difluoronitrobenzene, 3,4-difluoro-nitrobenzene
PubChem CID 123053
IUPAC Name 1,2-difluoro-4-nitrobenzene
SMILES C1=CC(=C(C=C1[N+](=O)[O-])F)F

Spezifikation

CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Density 1.4410g/mL
Boiling Point 80°C to 81°C (14.0 mmHg)
Flash Point 80°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula F2(C6H3)NO2
Refractive Index 1.5080 to 1.51
Quantity 100 mL
Beilstein 05, III, 609
Specific Gravity 1.441
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
Formula Weight 159.09
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3, 4-Difluoronitrobenzene
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • 3,4-Difluoronitrobenzene
Signalwort
  • Warning
Gefahrenkategorie
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Sicherheitshinweis
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.