missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluoroiodobenzene, 99%, Thermo Scientific™

Product Code. 10338610 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
25g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10338610 25g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10338610 Supplier Thermo Scientific Acros Supplier No. 233180250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 64248-58-4
CAS Min % 98.5
CAS Max % 100.0
Density 1.9900g/mL
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H3F2I
Linear Formula IC6H3F2
Refractive Index 1.5570 to 1.559
MDL Number MFCD00044630
Quantity 25g
Specific Gravity 1.99
Synonym 3,4-difluoroiodobenzene, 1,2-difluoro-4-iodo-benzene, benzene, 1,2-difluoro-4-iodo, 3,4-difluoro iodobenzene, pubchem3461, 3,4-difluoro-iodobenzene, 3,4-difluorophenyl iodide, 3,4-difluoro-1-iodobenzene, 1-iodo-3,4-difluoro-benzene
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
InChI Key KSASJELKLBIMSG-UHFFFAOYSA-N
SMILES FC1=C(F)C=C(I)C=C1
IUPAC Name 1,2-difluoro-4-iodobenzene
Molecular Weight (g/mol) 239.99
PubChem CID 2724444
Formula Weight 239.99
Percent Purity 99%
Chemical Name or Material 3, 4-Difluoroiodobenzene, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Difluoroiodobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.