missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorobenzylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Product Code. 15495325
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15495325 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15495325 Supplier Thermo Scientific Alfa Aesar Supplier No. H58167.AD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 307496-34-0 | C7H5BrF2Zn | 272.398 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Difluorobenzylzinc bromide is used as pharmaceutical intermediates.

Solubility
Reacts with water.

Notes
Store at 4°C. Incompatible with oxidizing agents, acid chlorides. Store under dry iner
TRUSTED_SUSTAINABILITY

Specifications

CAS 307496-34-0
Density 0.98
Molecular Formula C7H5BrF2Zn
MDL Number MFCD01311420
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive; May form a precipitate
Synonym zinc 2+ ion 3,4-difluorophenyl methanide bromide
Solubility Information Reacts with water.
InChI Key NXKQWSPHMLPJIC-UHFFFAOYSA-M
SMILES [CH2-]C1=CC(=C(C=C1)F)F.[Zn+2].[Br-]
Molecular Weight (g/mol) 272.398
PubChem CID 57371133
Formula Weight 272.4
Chemical Name or Material 3,4-Difluorobenzylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.