missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorobenzylamine, 95%, Thermo Scientific™

Product Code. 10416812 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10416812 5 g Glass bottle 5g
10013700 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10416812 Supplier Thermo Scientific Acros Supplier No. 311530050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Chemical Identifiers

CAS 72235-53-1
Molecular Formula C7H7F2N
Molecular Weight (g/mol) 143.14
MDL Number MFCD00010145
InChI Key PHLZUDXEBCQHKM-UHFFFAOYSA-N
PubChem CID 123572
IUPAC Name (3,4-difluorophenyl)methanamine
SMILES C1=CC(=C(C=C1CN)F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2100g/mL
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula F2C6H3CH2NH2
Refractive Index 1.4920 to 1.4940
Quantity 5 g
Specific Gravity 1.21
Synonym 3,4-difluorobenzylamine, 3,4-difluorophenyl methanamine, 3,4-difluorobenzyl amine, benzenemethanamine, 3,4-difluoro, 1-3,4-difluorophenyl methanamine, 3,4-difluorophenyl methylamine, 3, 4-difluorobenzylamine, pubchem1918, acmc-209omb, 3,4-difluoro-benzylamine
Formula Weight 143.14
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3, 4-Difluorobenzylamine, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.