missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorobenzyl chloride, 98%, Thermo Scientific™

Product Code. 11484267
Change view
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484267 2.5 g 2.5g
11494267 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11484267 Supplier Thermo Scientific Alfa Aesar Supplier No. B20367.88

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 698-80-6
Molecular Formula C7H5ClF2
Molecular Weight (g/mol) 162.56
MDL Number MFCD00236218
InChI Key VTGRVYJPIVZZGS-UHFFFAOYSA-N
Synonym 3,4-difluorobenzyl chloride, 4-chloromethyl-1,2-difluorobenzene, 3,4-difluorobenzylchloride, benzene, 4-chloromethyl-1,2-difluoro, 1-chloromethyl-3,4-difluorobenzene, alpha-chloro-3,4-difluorotoluene, 3, 4-difluorobenzyl chloride, pubchem7357, 4-chloromethyl-1,2-difluoro-benzene
PubChem CID 2736954
IUPAC Name 4-(chloromethyl)-1,2-difluorobenzene
SMILES FC1=CC=C(CCl)C=C1F

Specifications

Boiling Point 27°C to 29°C (0.4 mmHg)
Quantity 2.5 g
UN Number UN1760
Formula Weight 162.57
Percent Purity 98%
Chemical Name or Material 3,4-Difluorobenzyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.