Learn More
Thermo Scientific Maybridge 3,4-Difluorobenzaldehyde, 25g, 97%
Brand: Thermo Scientific Maybridge SB01773EB
3,4-Difluorobenzaldehyde, 25g, 97%
Code : 31
Additional Details : CAS Number : 34036-07-2 Weight : 0.07500kg
Brand: Thermo Scientific Maybridge SB01773EB
3,4-Difluorobenzaldehyde, 25g, 97%
Code : 31
Additional Details : CAS Number : 34036-07-2 Weight : 0.07500kg