missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluoroanisole, 98%

Product Code. 11434084
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11434084 25 g 25g
11424084 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11434084 Supplier Thermo Scientific Alfa Aesar Supplier No. A12474.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 115144-40-6 | C7H6F2O | 144.121 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115144-40-6
Molecular Formula C7H6F2O
Molecular Weight (g/mol) 144.121
MDL Number MFCD00010730
InChI Key DSEVNUCNUTYYHW-UHFFFAOYSA-N
Synonym 3,4-difluoroanisole, 1,2-difluoro-4-methoxy-benzene, benzene, 1,2-difluoro-4-methoxy, 3,4-difluoro-1-methoxybenzene, 1,2-difluor-4-methoxybenzol, pubchem3457, 3,4-difluoro-anisole, difluorobenzene,1-methoxy, acmc-2099pe
PubChem CID 568250
IUPAC Name 1,2-difluoro-4-methoxybenzene
SMILES COC1=CC(=C(C=C1)F)F

Specifications

Density 1.214
Flash Point 55°C (131°F)
Refractive Index 1.468
Quantity 25 g
UN Number UN1993
Beilstein 6717610
Formula Weight 144.12
Percent Purity 98%
Chemical Name or Material 3,4-Difluoroanisole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.