missing translation for 'onlineSavingsMsg'
Learn More

3',4'-Difluoroacetophenone, 98%

Product Code. 11402518
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11402518

Brand: Thermo Scientific Alfa Aesar A15832.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 369-33-5 | C8H6F2O | 156.132 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 369-33-5
Molecular Formula C8H6F2O
Molecular Weight (g/mol) 156.132
MDL Number MFCD00009891
InChI Key VWJSSJFLXRMYNV-UHFFFAOYSA-N
Synonym 3',4'-difluoroacetophenone, 1-3,4-difluorophenyl ethanone, 3,4-difluoroacetophenone, 1-3,4-difluorophenyl ethan-1-one, ethanone, 1-3,4-difluorophenyl, 1-3,4-difluoro-phenyl-ethanone, 3,4,-difluoroacetophenone, 1-acetyl-3,4-difluorobenzene, pubchem3089, acmc-209ipo
PubChem CID 123052
IUPAC Name 1-(3,4-difluorophenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1)F)F

Specifications

Melting Point 19°C to 20°C
Density 1.246
Boiling Point 94°C to 95°C (13 mmHg)
Flash Point 75°C (167°F)
Odor Characteristic
Refractive Index 1.4916
Quantity 5 g
Beilstein 2206856
Formula Weight 156.13
Percent Purity 98%
Chemical Name or Material 3',4'-Difluoroacetophenone
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.