missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluoro-2-methylbenzonitrile, 98%, Thermo Scientific™

Product Code. 15424147
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15424147 1 g 1g
15434147 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15424147 Supplier Thermo Scientific Alfa Aesar Supplier No. H31548.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 847502-83-4
Molecular Formula C8H5F2N
Molecular Weight (g/mol) 153.132
MDL Number MFCD07777154
InChI Key LMGHUTIUVKVZNH-UHFFFAOYSA-N
PubChem CID 24820441
IUPAC Name 3,4-difluoro-2-methylbenzonitrile
SMILES CC1=C(C=CC(=C1F)F)C#N

Specifica

Melting Point 39°C to 44°C
Quantity 1 g
UN Number UN3439
Synonym benzonitrile, 3,4-difluoro-2-methyl, 3,4-difluoro-o-tolunitrile, benzonitrile,3,4-difluoro-2-methyl, 2,3-difluoro-6-cyano-toluene, 3,4-difluoro-2-methyl benzonitrile, pubchem4769, 3,4-difluoro-2-methyl-benzonitrile
Formula Weight 153.13
Percent Purity 98%
Chemical Name or Material 3,4-Difluoro-2-methylbenzonitrile
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.