missing translation for 'onlineSavingsMsg'
Learn More

3,4-Diethoxybenzoic acid, 99%, Thermo Scientific™

Product Code. 10246500 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10246500 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10246500 Supplier Thermo Scientific Acros Supplier No. 206170250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 5409-31-4
CAS Min % 98.5
CAS Max % 100.0
Melting Point 167°C to 170°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C11H14O4
Linear Formula (C2H5O)2C6H3CO2H
MDL Number MFCD00002504
Quantity 25 g
Beilstein 10, 395
Synonym benzoic acid, 3,4-diethoxy, benzoicacid34diethoxy, 3,4-diethoxy-benzoic acid, acmc-1apyn, ksc271i5p, 3,4-bis ethyloxy benzoic acid, 3,4-diethoxybenzoic acid
InChI Key VVKCVAPLTRZJHH-UHFFFAOYSA-N
SMILES CCOC1=C(C=C(C=C1)C(=O)O)OCC
IUPAC Name 3,4-diethoxybenzoic acid
Molecular Weight (g/mol) 210.23
PubChem CID 79417
Formula Weight 210.23
Percent Purity 99%
Physical Form Amorphous Powder
Chemical Name or Material 3, 4-Diethoxybenzoic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Diethoxybenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.