missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorotoluene, TRC

Product Code. 30385717
Change view
Click to view available options
Quantity:
5 g
25 g
Product Code. Quantity
30385717 5 g
30372806 25 g
2 options
This item is not returnable. View return policy

Product Code. 30385717

Brand: Toronto Research Chemicals TRCD4367555G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-75-0
Formula Weight 159.9847
InChI Formula InChI=1S/C7H6Cl2/c1-5-2-3-6(8)7(9)4-5/h2-4H,1H3
IUPAC Name 1,2-dichloro-4-methylbenzene
Molecular Formula C7 H6 Cl2
Molecular Weight (g/mol) 161.03
SMILES Cc1ccc(Cl)c(Cl)c1
Synonym Toluene, 3,4-dichloro- (8CI), 1,2-Dichloro-4-methylbenzene, 3,4-Dichlorotoluene, NSC 8765
Chemical Name or Material 3,4-Dichlorotoluene

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.