missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorotoluene, 98%

Product Code. 11419662
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419662 1000 g 1000g
11429662 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419662 Supplier Thermo Scientific Alfa Aesar Supplier No. A10763.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95-75-0 | C7H6Cl2 | 161.025 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-75-0
Molecular Formula C7H6Cl2
Molecular Weight (g/mol) 161.025
MDL Number MFCD00000556
InChI Key WYUIWKFIFOJVKW-UHFFFAOYSA-N
Synonym 3,4-dichlorotoluene, benzene, 1,2-dichloro-4-methyl, toluene, 3,4-dichloro, 3,4-dichloro-toluene, ksc486q9r, chembl47525, benzene,2-dichloro-4-methyl, 1,2-dichloro-4-methyl-benzene, #
PubChem CID 7256
IUPAC Name 1,2-dichloro-4-methylbenzene
SMILES CC1=CC(=C(C=C1)Cl)Cl

Specifications

Melting Point ∼-15°C
Density 1.251
Boiling Point 200°C to 202°C
Flash Point 85°C (185°F)
Refractive Index 1.547
Quantity 1000 g
UN Number UN3082
Beilstein 1931687
Formula Weight 161.03
Percent Purity 98%
Chemical Name or Material 3,4-Dichlorotoluene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.