missing translation for 'onlineSavingsMsg'
Learn More

3',4'-Dichloropropiophenone, 98%

Product Code. 11483018
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11483018 25 g 25g
11473018 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11483018 Supplier Thermo Scientific Alfa Aesar Supplier No. A18740.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6582-42-9 | C9H8Cl2O | 203.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6582-42-9
Molecular Formula C9H8Cl2O
Molecular Weight (g/mol) 203.06
MDL Number MFCD00000554
InChI Key AJUAASWQUWIMHM-UHFFFAOYSA-N
Synonym 3',4'-dichloropropiophenone, 1-3,4-dichlorophenyl propan-1-one, 1-3,4-dichlorophenyl-1-propanone, 1-propanone, 1-3,4-dichlorophenyl, 1-propanone,1-3,4-dichlorophenyl, enamine_003892, acmc-1b5az, 3,4-dichloro propiophenone, 1-3,4-dichlorophenyl-1-propanone #
PubChem CID 599598
IUPAC Name 1-(3,4-dichlorophenyl)propan-1-one
SMILES CC(=O)CC1=CC=C(Cl)C(Cl)=C1

Specifications

Melting Point 44°C to 46°C
Boiling Point 136°C to 140°C (18 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Formula Weight 203.07
Percent Purity 98%
Chemical Name or Material 3',4'-Dichloropropiophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.