missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorophenylhydrazine hydrochloride, 98+%

Product Code. 11471618
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11471618 250 g 250g
11451618 10 g 10g
11461618 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11471618 Supplier Thermo Scientific Alfa Aesar Supplier No. A13906.30
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 19763-90-7 | C6H7Cl3N2 | 213.486 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19763-90-7
Molecular Formula C6H7Cl3N2
Molecular Weight (g/mol) 213.486
MDL Number MFCD00012937
InChI Key WIKZRYSGQPZTMM-UHFFFAOYSA-N
Synonym 3,4-dichlorophenylhydrazine hydrochloride, 3,4-dichlorophenyl hydrazine hydrochloride, 1-3,4-dichlorophenyl hydrazine hydrochloride, 3,4-dichlorophenyl hydrazine monohydrochloride, hydrazine, 3,4-dichlorophenyl-, monohydrochloride, 3,4-dichlorophenylhydrazinehydrochloride, pubchem7530, acmc-209f1i, ksc491e2j
PubChem CID 3084323
IUPAC Name (3,4-dichlorophenyl)hydrazine;hydrochloride
SMILES C1=CC(=C(C=C1NN)Cl)Cl.Cl

Specifications

Melting Point ∼229°C (decomposition)
Quantity 250 g
Beilstein 3703934
Formula Weight 213.5
Percent Purity ≥98%
Chemical Name or Material 3,4-Dichlorophenylhydrazine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.