missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorophenylacetonitrile, 98%

Product Code. 11460203
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460203 25 g 25g
11470203 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11460203 Supplier Thermo Scientific Alfa Aesar Supplier No. A11382.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3218-49-3 | C8H5Cl2N | 186.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3218-49-3
Molecular Formula C8H5Cl2N
Molecular Weight (g/mol) 186.04
MDL Number MFCD00001909
InChI Key QWZNCAFWRZZJMA-UHFFFAOYSA-N
Synonym 3,4-dichlorophenylacetonitrile, 2-3,4-dichlorophenyl acetonitrile, 3,4-dichlorobenzyl cyanide, 3,4-dichlorophenyl acetonitrile, benzeneacetonitrile, 3,4-dichloro, 3,4-dichlorobenzeneacetonitrile, 2-3,4-dichlorophenyl ethanenitrile, 3,4-dichlorphenyl acetonitrile, pubchem16506, 3,4-dichlorobenzylcyanide
PubChem CID 76690
IUPAC Name 2-(3,4-dichlorophenyl)acetonitrile
SMILES ClC1=CC=C(CC#N)C=C1Cl

Specifications

Melting Point 38°C to 42°C
Boiling Point 169°C to 171°C (12 mmHg)
Flash Point >110°C (230°F)
Odor Characteristic
Quantity 25 g
UN Number UN3439
Beilstein 638749
Formula Weight 186.04
Percent Purity 98%
Chemical Name or Material 3,4-Dichlorophenylacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.