missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorophenylacetic acid, 98%

Product Code. 11467567
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11467567 5 g 5g
11477567 25 g 25g
11487567 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11467567 Supplier Thermo Scientific Alfa Aesar Supplier No. A15653.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5807-30-7 | C8H6Cl2O2 | 205.034 g/mol

It is used in the parallel solid phase synthesis of tetra substituted diethylenetriamines via selective amide alkylation and exhaustive reduction of N-acylated dipeptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the parallel solid phase synthesis of tetra substituted diethylenetriamines via selective amide alkylation and exhaustive reduction of N-acylated dipeptides.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5807-30-7
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.034
MDL Number MFCD00004333
InChI Key ZOUPGSMSNQLUNW-UHFFFAOYSA-N
Synonym 3,4-dichlorophenylacetic acid, 2-3,4-dichlorophenyl acetic acid, 3,4-dichlorophenyl acetic acid, 3,4-dichlorobenzene acetic acid, benzeneacetic acid, 3,4-dichloro, pubchem20195, acmc-209m2g, 3,4dichlorophenylacetic acid, 3,4-dichlorophenylaceticacid
PubChem CID 79874
IUPAC Name 2-(3,4-dichlorophenyl)acetic acid
SMILES C1=CC(=C(C=C1CC(=O)O)Cl)Cl

Specifications

Melting Point 87°C to 92°C
Odor Odorless
Quantity 5 g
Beilstein 1953781
Solubility Information Insoluble in water.
Formula Weight 205.04
Percent Purity 98%
Chemical Name or Material 3,4-Dichlorophenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.