missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorophenylacetic acid, 98%, Thermo Scientific™

Product Code. 10592261 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10592261 25 g Glass bottle 25g
10241093 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10592261 Supplier Thermo Scientific Acros Supplier No. 279100250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5807-30-7
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.034
MDL Number MFCD00004333
InChI Key ZOUPGSMSNQLUNW-UHFFFAOYSA-N
Synonym 3,4-dichlorophenylacetic acid, 2-3,4-dichlorophenyl acetic acid, 3,4-dichlorophenyl acetic acid, 3,4-dichlorobenzene acetic acid, benzeneacetic acid, 3,4-dichloro, pubchem20195, acmc-209m2g, 3,4dichlorophenylacetic acid, 3,4-dichlorophenylaceticacid
PubChem CID 79874
IUPAC Name 2-(3,4-dichlorophenyl)acetic acid
SMILES C1=CC(=C(C=C1CC(=O)O)Cl)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 86°C to 90°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula ClC6H3CH2CO2H
Quantity 25 g
Formula Weight 205.04
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 3, 4-Dichlorophenylacetic acid, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.