missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorophenol, 99%

Product Code. 10107523 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10107523 50 g Glass bottle 50g
10497290 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10107523 Supplier Thermo Scientific Acros Supplier No. 113600500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 95-77-2 | C6H4Cl2O | 163.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-77-2
Molecular Formula C6H4Cl2O
Molecular Weight (g/mol) 163.00
MDL Number MFCD00002258
InChI Key WDNBURPWRNALGP-UHFFFAOYSA-N
Synonym 4,5-dichlorophenol, phenol, 3,4-dichloro, 3,3-dichloro-hydroxybenzene, unii-070ftm6dvf, ccris 5904, 070ftm6dvf, 3,4-dcp, 3,4-dichlorphenol, 3,4dichlorophenol, 3,4 dichlorophenol
PubChem CID 7258
ChEBI CHEBI:34323
IUPAC Name 3,4-dichlorophenol
SMILES OC1=CC=C(Cl)C(Cl)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 65°C to 68°C
Color Brown to White
Boiling Point 145°C to 146°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula Cl2C6H3OH
Quantity 50 g
Beilstein 06, 190
Solubility Information Solubility in water: slightly soluble
Formula Weight 163
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 3, 4-Dichlorophenol
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Dichlorophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.