missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorophenol, 97%

Product Code. 11474953
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474953 25 g 25g
11484953 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11474953 Supplier Thermo Scientific Alfa Aesar Supplier No. A12256.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95-77-2 | C6H4Cl2O | 163.00 g/mol

It is also a photo-degradation product of the common antibacterial and antifungal agent triclosan along with the dioxin 2,8-dichlorodibenzo-p-dioxin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is also a photo-degradation product of the common antibacterial and antifungal agent triclosan along with the dioxin 2,8-dichlorodibenzo-p-dioxin.

Solubility
Slightly soluble ion water 9.2 g/L at 20degC. Soluble in aqueous alkali, oxygenated and chlorinated solvents. Soluble in ethanol, benzene, chloroform and ethyl ether.

Notes
Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents, acid, acid fumes, acid chlorides, acid anhydrides, caustics. Keep container tightly closed. Store in an area without drain or sewer access. Separated from strong oxidants and food and feedstuffs.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-77-2
Molecular Formula C6H4Cl2O
Molecular Weight (g/mol) 163.00
MDL Number MFCD00002258
InChI Key WDNBURPWRNALGP-UHFFFAOYSA-N
PubChem CID 7258
ChEBI CHEBI:34323
IUPAC Name 3,4-dichlorophenol
SMILES OC1=CC=C(Cl)C(Cl)=C1

Specifications

Melting Point 64°C to 68°C
Boiling Point 252°C to 255°C
Odor Phenol-like
Quantity 25 g
UN Number UN2020
Beilstein 1907693
Synonym 4,5-dichlorophenol, phenol, 3,4-dichloro, 3,3-dichloro-hydroxybenzene, unii-070ftm6dvf, ccris 5904, 070ftm6dvf, 3,4-dcp, 3,4-dichlorphenol, 3,4dichlorophenol, 3,4 dichlorophenol
Solubility Information Slightly soluble ion water 9.2g/L at 20degC. Soluble in aqueous alkali,oxygenated and chlorinated solvents. Soluble in ethanol,benzene,chloroform and ethyl ether.
Formula Weight 163
Percent Purity 99%
Chemical Name or Material 3,4-Dichlorophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.