missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dichloronitrobenzene, 95%, Thermo Scientific™
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Specifications
Specifications
CAS Min % | 94.0 |
CAS Max % | 100.0 |
Melting Point | 39°C to 45°C |
Color | Yellow to Brown |
Infrared Spectrum | Authentic |
Assay Percent Range | 95% |
Packaging | Glass bottle |
Linear Formula | Cl2C6H3NO2 |
Quantity | 250 g |
Beilstein | 05, 246 |
Show More |
Safety and Handling
Product Identifier
- 3,4-Dichloronitrobenzene
- Warning
- Acute toxicity Category 4
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'