missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzyl isothiocyanate, 97%, Thermo Scientific™

Product Code. 11342577
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11342577 5 g 5g
11332577 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11342577 Supplier Thermo Scientific Alfa Aesar Supplier No. L11701.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18967-42-5
Molecular Formula C8H5Cl2NS
Molecular Weight (g/mol) 218.095
MDL Number MFCD00041113
InChI Key DPSSHXWTNIGVMQ-UHFFFAOYSA-N
Synonym 3,4-dichlorobenzyl isothiocyanate, 1,2-dichloro-4-isothiocyanatomethyl benzene, isothiocyanic acid, 3,4-dichlorobenzyl ester, benzene, 1,2-dichloro-4-isothiocyanatomethyl, acmc-1bqjy, wln: scn1r cg dg, 4-12-00-02393 beilstein handbook reference, 3,4-dichlorobenzylisothiocyanate, 3,4-dichlorophenyl methanisothiocyanate, isothiocyanic acid,4-dichlorobenzyl ester
PubChem CID 29350
IUPAC Name 1,2-dichloro-4-(isothiocyanatomethyl)benzene
SMILES C1=CC(=C(C=C1CN=C=S)Cl)Cl

Specifications

Melting Point 26°C to 28°C
Density 1.38
Boiling Point 130°C (1 mmHg)
Odor Irritating
Refractive Index 1.6295
Quantity 5 g
UN Number UN2923
Beilstein 2937608
Sensitivity Moisture sensitive
Formula Weight 218.11
Percent Purity 97%
Chemical Name or Material 3,4-Dichlorobenzyl isothiocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.