missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzyl Chloride, TRC

Product Code. 30006877
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006877 10 g 10g
30005254 5 g 5g
30006017 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006877 Supplier Toronto Research Chemicals Supplier No. TRCD43841510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-47-6
Molecular Formula C7 H5 Cl3
Molecular Weight (g/mol) 195.47
Synonym alpha,3,4-Trichlorotoluene, Benzene, 1,2-dichloro-4-(chloromethyl)-, Toluene, α,3,4-trichloro- (7CI,8CI), 1,2-Dichloro-4-(chloromethyl)benzene, 3,4,α-Trichlorotoluene, 3,4-Dichloro-1-(chloromethyl)benzene, 3,4-Dichlorobenzyl chloride, 3,4-Dichlorophenylmethyl chloride, 4-Chloromethyl-1,2-dichlorobenzene, NSC 406893, α,3,4-Trichlorotoluene
IUPAC Name 1,2-dichloro-4-(chloromethyl)benzene
SMILES ClCc1ccc(Cl)c(Cl)c1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C7H5Cl3/c8-4-5-1-2-6(9)7(10)3-5/h1-3H,4H2
Formula Weight 193.9457
Chemical Name or Material alpha,3,4-Trichlorotoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.