missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzyl chloride, 98%

Product Code. 11413258
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
250 g
Unit Size:
100g
250g
25g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413258 25 g 25g
11399529 50 g 50g
11423258 100 g 100g
11309539 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11413258 Supplier Thermo Scientific Alfa Aesar Supplier No. A17672.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 102-47-6 | C7H5Cl3 | 195.467 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-47-6
Molecular Formula C7H5Cl3
Molecular Weight (g/mol) 195.467
MDL Number MFCD00000906
InChI Key YZIFVWOCPGPNHB-UHFFFAOYSA-N
Synonym 3,4-dichlorobenzyl chloride, 1,2-dichloro-4-chloromethyl benzene, alpha,3,4-trichlorotoluene, benzene, 1,2-dichloro-4-chloromethyl, unii-451pb8smie, 1,2-dichloro-4-chloromethyl-benzene, 3,4-dichlorobenzylchloride, 451pb8smie, 3,4-dichlorophenylmethyl chloride, toluene, .alpha.,3,4-trichloro
PubChem CID 7609
IUPAC Name 1,2-dichloro-4-(chloromethyl)benzene
SMILES C1=CC(=C(C=C1CCl)Cl)Cl

Specifications

Melting Point -3°C
Density 1.416
Boiling Point 99°C to 101°C (2 mmHg)
Flash Point 155°C (311°F)
Odor Pungent
Refractive Index 1.5766
Quantity 25 g
UN Number UN3265
Beilstein 386644
Formula Weight 195.48
Percent Purity 98%
Chemical Name or Material 3,4-Dichlorobenzyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.