missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzyl bromide, 98%

Product Code. 11347365
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347365 5 g 5g
11357365 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347365 Supplier Thermo Scientific Alfa Aesar Supplier No. L05254.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18880-04-1 | C7H5BrCl2 | 239.921 g/mol

3,4-Dichlorobenzyl bromide is used as a pharmaceutical intermediate. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dichlorobenzyl bromide is used as a pharmaceutical intermediate. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18880-04-1
Molecular Formula C7H5BrCl2
Molecular Weight (g/mol) 239.921
MDL Number MFCD00041363
InChI Key XLWSBDFQAJXCQX-UHFFFAOYSA-N
Synonym 3,4-dichlorobenzyl bromide, 4-bromomethyl-1,2-dichlorobenzene, benzene, 4-bromomethyl-1,2-dichloro, 3,4-dichlorobenzylbromide, alpha-bromo-3,4-dichlorotoluene, acmc-209erq, 3,4 dichlorobenzyl bromide, 3,4-dichloro-benzyl bromide, ksc493a6b, timtec-bb sbb005763
PubChem CID 639726
IUPAC Name 4-(bromomethyl)-1,2-dichlorobenzene
SMILES C1=CC(=C(C=C1CBr)Cl)Cl

Specifications

Density 1.65
Boiling Point 116°C to 119°C (1 mmHg)
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.6065
Quantity 5 g
UN Number UN3265
Beilstein 742579
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 239.93
Percent Purity 98%
Chemical Name or Material 3,4-Dichlorobenzyl bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.