missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzyl bromide, 97%

Product Code. 10308890 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10308890 25 g Glass bottle 25g
10150991 100 g Glass bottle 100g
10185353 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10308890 Supplier Thermo Scientific Acros Supplier No. 326160250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 18880-04-1 | C7H5BrCl2 | 239.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18880-04-1
Molecular Formula C7H5BrCl2
Molecular Weight (g/mol) 239.93
MDL Number MFCD00041363
InChI Key XLWSBDFQAJXCQX-UHFFFAOYSA-N
Synonym 3,4-dichlorobenzyl bromide, 4-bromomethyl-1,2-dichlorobenzene, benzene, 4-bromomethyl-1,2-dichloro, 3,4-dichlorobenzylbromide, alpha-bromo-3,4-dichlorotoluene, acmc-209erq, 3,4 dichlorobenzyl bromide, 3,4-dichloro-benzyl bromide, ksc493a6b, timtec-bb sbb005763
PubChem CID 639726
IUPAC Name 4-(bromomethyl)-1,2-dichlorobenzene
SMILES C1=CC(=C(C=C1CBr)Cl)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Brown
Density 1.6500g/mL
Boiling Point 115°C to 120°C (1.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.6050 to 1.607
Quantity 25 g
Specific Gravity 1.65
Formula Weight 239.93
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3, 4-Dichlorobenzyl bromide
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Dichlorobenzyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.