missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzoyl Chloride, TRC

Product Code. 30386336
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386336 50 g 50g
30376945 10 g 10g
30388211 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386336 Supplier Toronto Research Chemicals Supplier No. TRCD43173050G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3024-72-4
Molecular Formula C7H3Cl3O
Molecular Weight (g/mol) 209.46
Synonym 3,4-Dichloro-benzoyl Chloride
IUPAC Name 3,4-dichlorobenzoyl chloride
SMILES ClC(=O)c1ccc(Cl)c(Cl)c1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C7H3Cl3O/c8-5-2-1-4(7(10)11)3-6(5)9/h1-3H
Formula Weight 207.9249
Chemical Name or Material 3,4-Dichlorobenzoyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.