missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzoyl chloride, 97%

Product Code. 11468856
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11468856 100 g 100g
11458856 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11468856 Supplier Thermo Scientific Alfa Aesar Supplier No. A17006.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3024-72-4 | C7H3Cl3O | 209.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3024-72-4
Molecular Formula C7H3Cl3O
Molecular Weight (g/mol) 209.45
MDL Number MFCD00000672
InChI Key VTXNOVCTHUBABW-UHFFFAOYSA-N
PubChem CID 76403
IUPAC Name 3,4-dichlorobenzoyl chloride
SMILES C1=CC(=C(C=C1C(=O)Cl)Cl)Cl

Specifications

Melting Point 30°C to 34°C
Boiling Point 242°C
Flash Point 142°C (287°F)
Quantity 100 g
UN Number UN3261
Beilstein 607485
Sensitivity Moisture sensitive
Synonym benzoyl chloride, 3,4-dichloro, 3,4-dichlorobenzoylchloride, 3,4-dichloro benzoyl chloride, 2,3-dichlorobenzoyl, 3,4dichlorobenzoyl chloride, acmc-1aj45, 3,4 dichlorobenzoyl chloride, 3,4-dichlorobenzoyl-chloride, ksc225o5d, 3,4-dichloro-benzoyl chloride
Formula Weight 209.46
Percent Purity 97%
Chemical Name or Material 3,4-Dichlorobenzoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.