missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzoyl chloride, 97%

Product Code. 10255150 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10255150 100 g Glass Bottle 100g
10041510 5 g Glass Bottle 5g
10448300 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10255150 Supplier Thermo Scientific Acros Supplier No. 150231000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3024-72-4 | C7H3Cl3O | 209.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3024-72-4
Molecular Formula C7H3Cl3O
Molecular Weight (g/mol) 209.45
MDL Number MFCD00000672
InChI Key VTXNOVCTHUBABW-UHFFFAOYSA-N
PubChem CID 76403
IUPAC Name 3,4-dichlorobenzoyl chloride
SMILES C1=CC(=C(C=C1C(=O)Cl)Cl)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Linear Formula Cl2C6H3COCl
Quantity 100 g
Beilstein 09, 344
Synonym benzoyl chloride, 3,4-dichloro, 3,4-dichlorobenzoylchloride, 3,4-dichloro benzoyl chloride, 2,3-dichlorobenzoyl, 3,4dichlorobenzoyl chloride, acmc-1aj45, 3,4 dichlorobenzoyl chloride, 3,4-dichlorobenzoyl-chloride, ksc225o5d, 3,4-dichloro-benzoyl chloride
Formula Weight 209.46
Percent Purity 97%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 3, 4-Dichlorobenzoyl chloride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Dichlorobenzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.