missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzotrifluoride, 97%, Thermo Scientific™

Product Code. 11467407
Change view
Click to view available options
Quantity:
50 g
250 g
500 g
Unit Size:
250g
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11467407 50 g 50g
11319539 250 g 250g
11329539 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11467407 Supplier Thermo Scientific Alfa Aesar Supplier No. A15098.18

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 328-84-7
Molecular Formula C7H3Cl2F3
Molecular Weight (g/mol) 214.996
MDL Number MFCD00000555
InChI Key XILPLWOGHPSJBK-UHFFFAOYSA-N
Synonym 3,4-dichlorobenzotrifluoride, 1,2-dichloro-4-trifluoromethyl benzene, benzene, 1,2-dichloro-4-trifluoromethyl, 3,4-dichloro-alpha,alpha,alpha-trifluorotoluene, 3,4-dichlorotrifluoromethylbenzene, unii-7g60xcu4gg, 3,4-dichlorophenyltrifluoromethane, 7g60xcu4gg, 3,4-dichloro-1-trifluoromethyl benzene, 3,4-dcbte
PubChem CID 9481
IUPAC Name 1,2-dichloro-4-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1C(F)(F)F)Cl)Cl

Specifications

Melting Point -12°C
Density 1.479
Boiling Point 173°C to 174°C
Flash Point 65°C (149°F)
Refractive Index 1.475
Quantity 50 g
UN Number UN1760
Beilstein 1950151
Formula Weight 215
Percent Purity 97%
Chemical Name or Material 3,4-Dichlorobenzotrifluoride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.