missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzophenone, TRC

Product Code. 30381282
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381282 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30381282 Supplier Toronto Research Chemicals Supplier No. TRCD4319651G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,4-Dichlorobenzophenone
CAS 6284-79-3
Molecular Formula C13H8Cl2O
Quantity 1 g
Synonym Benzophenone, 3,4-dichloro- (6CI,8CI), (3,4-Dichlorophenyl)phenylmethanone, (3,4-Dichlorophenyl)(phenyl)methanone, 3,4-Dichlorophenyl phenyl ketone, NSC 5249, Phenyl 3,4-dichlorophenyl ketone
InChI Formula InChI=1S/C13H8Cl2O/c14-11-7-6-10(8-12(11)15)13(16)9-4-2-1-3-5-9/h1-8H
SMILES Clc1ccc(cc1Cl)C(=O)c2ccccc2
IUPAC Name (3,4-dichlorophenyl)-phenylmethanone
Molecular Weight (g/mol) 251.11
Formula Weight 249.9952
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.