missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 11411978
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11411978

Brand: Thermo Scientific Alfa Aesar A14576.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-31-7
Molecular Formula C6H3Cl3O2S
Molecular Weight (g/mol) 245.50
MDL Number MFCD00041255
InChI Key NYIBPWGZGSXURD-UHFFFAOYSA-N
Synonym 3,4-dichlorobenzene-1-sulfonyl chloride, 3,4-dichlorophenylsulfonyl chloride, 3,4-dichlorobenzenesulphonyl chloride, benzenesulfonyl chloride, 3,4-dichloro, 3,4-dichlorobenzenesulfonylchloride, 3,4-dichloro-benzenesulfonyl chloride, 3,4-dichlorobenzene-sulfonyl chloride, 3,4-dichloro benzene sulfonyl chloride, 3,4-dichlorophenyl chlorosulfone, pubchem5118
PubChem CID 7383
IUPAC Name 3,4-dichlorobenzenesulfonyl chloride
SMILES ClC1=CC=C(C=C1Cl)S(Cl)(=O)=O

Specifications

Melting Point 20°C to 22°C
Density 1.572
Boiling Point 253°C
Flash Point >110°C (230°F)
Refractive Index 1.589
Quantity 25 g
UN Number UN3265
Beilstein 1956417
Sensitivity Moisture sensitive
Formula Weight 245.51
Percent Purity 98%
Chemical Name or Material 3,4-Dichlorobenzenesulfonyl chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.