missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzeneboronic acid, 97%

Product Code. 11445707
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11445707 1 g 1g
11455707 5 g 5g
11465707 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11445707 Supplier Thermo Scientific Alfa Aesar Supplier No. B24292.03
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 151169-75-4 | C6H5BCl2O2 | 190.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151169-75-4
Molecular Formula C6H5BCl2O2
Molecular Weight (g/mol) 190.81
MDL Number MFCD01074646
InChI Key JKIGHOARKAIPJI-UHFFFAOYSA-N
Synonym 3,4-dichlorophenyl boronic acid, 3,4-dichlorobenzeneboronic acid, 3,4-dichlorophenylboronicacid, 3,4-dichlorophenyl boranediol, 3,4-dichlorophenylbornic acid, chembl20776, 3,4-dichloro benzene boronic acid, boronic acid, 3,4-dichlorophenyl, pubchem1812
PubChem CID 2734330
IUPAC Name (3,4-dichlorophenyl)boronic acid
SMILES OB(O)C1=CC=C(Cl)C(Cl)=C1

Specifications

Melting Point 270°C to 274°C
Quantity 1 g
Beilstein 7369790
Formula Weight 190.82
Percent Purity 97%
Chemical Name or Material 3,4-Dichlorobenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.