missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzaldehyde, 97%

Product Code. 11399859
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11399859 5 g 5g
11309869 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11399859 Supplier Thermo Scientific Alfa Aesar Supplier No. B24706.06
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6287-38-3 | C7H4Cl2O | 175.008 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6287-38-3
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.008
MDL Number MFCD00003351
InChI Key ZWUSBSHBFFPRNE-UHFFFAOYSA-N
Synonym benzaldehyde, 3,4-dichloro, unii-i6z632wsj5, ccris 6015, pubchem8230, 3,4-dichlorobenzaldehye, 3,4-dichlorobenzaldhyde, 3,4 dichlorobenzaldehyde, 3,4-dichlorobenzaidehyde, 4,3-dichlorobenzaldehyde, dsstox_cid_4971
PubChem CID 22710
IUPAC Name 3,4-dichlorobenzaldehyde
SMILES C1=CC(=C(C=C1C=O)Cl)Cl

Specifications

Melting Point 41°C to 44°C
Boiling Point 247°C to 248°C
Flash Point 135°C (275°F)
Quantity 5 g
UN Number UN1759
Beilstein 636379
Sensitivity Air Sensitive
Formula Weight 175.01
Percent Purity 97%
Chemical Name or Material 3,4-Dichlorobenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.