missing translation for 'onlineSavingsMsg'
Learn More

3',4'-Dichloroacetophenone, 99%, ACROS Organics™

Product Code. 10265890 Shop All Thermo Scientific Chemicals Products
Click to view available options
:
100g; Glass bottle
This item is not returnable. View return policy

Product Code. 10265890

Brand: Thermo Scientific Acros 170951000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 2642-63-9
CAS Min % 98.5
CAS Max % 100.0
Melting Point 72°C to 76°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C8H6Cl2O
Linear Formula Cl2C6H3COCH3
MDL Number MFCD00000553
Quantity 100 g
Beilstein 07, II, 219
Synonym 3',4'-dichloroacetophenone, 1-3,4-dichlorophenyl ethanone, 3,4-dichloroacetophenone, 1-3,4-dichlorophenyl ethan-1-one, ethanone, 1-3,4-dichlorophenyl, acetophenone, 3',4'-dichloro, 1-acetyl-3,4-dichlorobenzene, 3,4 dichloro acetophenone, pubchem3383, 3,4-dicloroacetophenone
InChI Key WBPAOUHWPONFEQ-UHFFFAOYSA-N
SMILES CC(=O)C1=CC=C(Cl)C(Cl)=C1
IUPAC Name 1-(3,4-dichlorophenyl)ethan-1-one
Molecular Weight (g/mol) 189.04
PubChem CID 75841
Formula Weight 189.04
Percent Purity ≥98.5%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 3';, 4';-Dichloroacetophenone, 99%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.