missing translation for 'onlineSavingsMsg'
Learn More

3',4'-Dichloroacetophenone, 98+%

Product Code. 11480074
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11480074

Brand: Thermo Scientific Alfa Aesar A13568.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2642-63-9 | C8H6Cl2O | 189.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2642-63-9
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.04
MDL Number MFCD00000553
InChI Key WBPAOUHWPONFEQ-UHFFFAOYSA-N
Synonym 3',4'-dichloroacetophenone, 1-3,4-dichlorophenyl ethanone, 3,4-dichloroacetophenone, 1-3,4-dichlorophenyl ethan-1-one, ethanone, 1-3,4-dichlorophenyl, acetophenone, 3',4'-dichloro, 1-acetyl-3,4-dichlorobenzene, 3,4 dichloro acetophenone, pubchem3383, 3,4-dicloroacetophenone
PubChem CID 75841
IUPAC Name 1-(3,4-dichlorophenyl)ethanone
SMILES CC(=O)C1=CC=C(Cl)C(Cl)=C1

Specifications

Melting Point 72°C to 76°C
Boiling Point 134°C to 135 mmHg) (12 mmHg)
Quantity 25 g
Beilstein 742534
Formula Weight 189.04
Percent Purity ≥98%
Chemical Name or Material 3',4'-Dichloroacetophenone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.