missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichloro-1,2,5-thiadiazole, Mikromol

Product Code. 30459969
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30459969 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30459969 Supplier Mikromol Supplier No. MM0440.230500
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3,4-Dichloro-1,2,5-thiadiazole, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,4-Dichloro-1,2,5-thiadiazole
CAS 5728-20-1
Analyte or Component Names 3,4-Dichloro-1,2,5-thiadiazole
Shipping Condition Room Temperature
Molecular Formula C2Cl2N2S
Certifications/Compliance ISO 9001
Synonym 4,5-Dichloro-2,1,3-thiadiazole;
Solution Type Neat
SMILES Clc1nsnc1Cl
Molecular Weight (g/mol) 155.01
Quantity 500 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.