missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichloro-1,2,5-thiadiazole, 99%

Product Code. 11439273
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439273 50 g 50g
11429273 10 g 10g
11449273 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439273 Supplier Thermo Scientific Alfa Aesar Supplier No. A11738.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5728-20-1 | C2Cl2N2S | 155.00 g/mol

3,4-Dichloro-1,2,5-thiadiazole is a chlorinated thiadiazole that is a potent inhibitors of nitrification in soil. 3,4-Dichloro-1,2,5-thiadiazole is used as a building block in the preparaton of the β-adrenergic blocking agent, Timolol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dichloro-1,2,5-thiadiazole is a chlorinated thiadiazole that is a potent inhibitors of nitrification in soil. 3,4-Dichloro-1,2,5-thiadiazole is used as a building block in the preparaton of the β-adrenergic blocking agent, Timolol.

Solubility
Soluble in chloroform and methanol.

Notes
Hygroscopic. Store under dry inert gas. Incompatible with moisture and oxidizing agents, acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5728-20-1
Molecular Formula C2Cl2N2S
Molecular Weight (g/mol) 155.00
MDL Number MFCD00010072
InChI Key YNZQOVYRCBAMEU-UHFFFAOYSA-N
Synonym 1,2,5-thiadiazole, 3,4-dichloro, dichloro-1,2,5-thiadiazole, 3,4-dichloro-1,2,5 thiadiazole, pubchem20790, acmc-1al9l, ksc272e3d, 4,5-dichloro-2,1,3-thiadiazole, 3,4-bis chloranyl-1,2,5-thiadiazole, 3,4-dichloro-1,2,5-thiadiazole
PubChem CID 79804
IUPAC Name 3,4-dichloro-1,2,5-thiadiazole
SMILES ClC1=NSN=C1Cl

Specifications

Density 1.644
Boiling Point 157°C to 159°C
Flash Point >100°C (212°F)
Refractive Index 1.562
Quantity 50 g
UN Number UN2810
Beilstein 606585
Sensitivity Hygroscopic
Solubility Information Soluble in chloroform and methanol.
Formula Weight 155.01
Percent Purity 99%
Chemical Name or Material 3,4-Dichloro-1,2,5-thiadiazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.