Learn More
3,4-Diaminothiophene dihydrochloride, 96%, Thermo Scientific Chemicals
£66.00 - £203.00
Chemical Identifiers
CAS | 90069-81-1 |
---|---|
Molecular Formula | C4H6N2S·2HCl |
Molecular Weight (g/mol) | 187.09 |
MDL Number | MFCD00671511 |
InChI Key | RAMOMCXNLLLICQ-UHFFFAOYSA-N |
Synonym | 3,4-diaminothiophene dihydrochloride, thiophene-3,4-diamine dihydrochloride, 3,4-diaminothiophenedihydrochloride, pubchem21822, acmc-209r4q, 3,4-diaminothiophene 2hcl, 3?4-diaminothiophene dihydrochloride, 3,4-thiophenediamine, dihydrochloride, thiophene-3,4-diamine-hydrogen chloride 1/2 |
PubChem CID | 12707342 |
IUPAC Name | thiophene-3,4-diamine;dihydrochloride |
SMILES | C1=C(C(=CS1)N)N.Cl.Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10315901
|
Thermo Scientific Acros
325890010 |
1 g | Glass bottle |
£66.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10389360
|
Thermo Scientific Acros
325890050 |
5 g | Glass bottle |
£203.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
90069-81-1 | |
187.09 | |
RAMOMCXNLLLICQ-UHFFFAOYSA-N | |
12707342 | |
C1=C(C(=CS1)N)N.Cl.Cl |
C4H6N2S·2HCl | |
MFCD00671511 | |
3,4-diaminothiophene dihydrochloride, thiophene-3,4-diamine dihydrochloride, 3,4-diaminothiophenedihydrochloride, pubchem21822, acmc-209r4q, 3,4-diaminothiophene 2hcl, 3?4-diaminothiophene dihydrochloride, 3,4-thiophenediamine, dihydrochloride, thiophene-3,4-diamine-hydrogen chloride 1/2 | |
thiophene-3,4-diamine;dihydrochloride |
Specifications
90069-81-1 | |
100.0 | |
Brown or Gray | |
Authentic | |
C4H6N2S·2HCl | |
3,4-diaminothiophene dihydrochloride, thiophene-3,4-diamine dihydrochloride, 3,4-diaminothiophenedihydrochloride, pubchem21822, acmc-209r4q, 3,4-diaminothiophene 2hcl, 3?4-diaminothiophene dihydrochloride, 3,4-thiophenediamine, dihydrochloride, thiophene-3,4-diamine-hydrogen chloride 1/2 | |
C1=C(C(=CS1)N)N.Cl.Cl | |
187.09 | |
187.09 | |
Crystalline Powder |
95.0 | |
240.0°C to 245.0°C | |
3% max. | |
95% min. (on dry substance) (Argentometry) | |
MFCD00671511 | |
RAMOMCXNLLLICQ-UHFFFAOYSA-N | |
thiophene-3,4-diamine;dihydrochloride | |
12707342 | |
96% | |
3, 4-Diaminothiophene dihydrochloride, 99% |
Safety and Handling
GHS H Statement
Toxic if swallowed.
May cause an allergic skin reaction.
Causes serious eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Danger
missing translation for 'tsca' : TSCA
RUO – Research Use Only