missing translation for 'onlineSavingsMsg'
Learn More

3,4-Di-O-acetyl-L-arabinal, 97%, Thermo Scientific™

Product Code. 10785673 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10785673 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10785673 Supplier Thermo Scientific Acros Supplier No. 370380010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 3945-18-4
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C9H12O5
Refractive Index 1.4580 to 1.46
MDL Number MFCD04112729,MFCD08704080
Quantity 1 g
Beilstein 17, IV, 2035
Synonym 3,4-di-o-acetyl-l-arabinal, 3s,4s-3-acetyloxy-3,4-dihydro-2h-pyran-4-yl acetate, d-di-o-acetylarabinal, di-o-acetyl-d-arabinal, 3,4-di-o-acetyl-d-ribal, 3,4-dihydro-2h-pyran-3,4-diyl diacetate, 1,2-dideoxy-d-erythro-pent-1-enopyranose diacetate, 3s,4s-3,4-dihydro-2h-pyran-3,4-diyl diacetate, 3s,4s-4-acetyloxy-3,4-dihydro-2h-pyran-3-yl acetate, 1,5-anhydro-2-deoxy-d-erythro-pent-1-enitol 3,4-diacetate
InChI Key OWKCFBYWQGPLSJ-BDAKNGLRSA-N
SMILES CC(=O)O[C@H]1COC=C[C@H]1OC(C)=O
IUPAC Name [(3R,4S)-4-acetyloxy-3,4-dihydro-2H-pyran-3-yl] acetate
Molecular Weight (g/mol) 200.19
PubChem CID 8030588
Formula Weight 200.19
Percent Purity 97%
Physical Form Oil
Chemical Name or Material 3, 4-Di-O-acetyl-L-arabinal
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.