missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Cyanophenyl)propionic acid, 96%

Product Code. 15437237
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15437237 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15437237 Supplier Thermo Scientific Alfa Aesar Supplier No. H34297.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 42287-94-5 | C10H9NO2 | 175.187 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42287-94-5
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD08234823
InChI Key RHIGOXBTMPLABF-UHFFFAOYSA-N
Synonym 3-4-cyanophenyl propanoic acid, 3-4-cyano-phenyl-propionic acid, 3-4-cyanophenyl-propionic acid, 4-cyanobenzenepropanoic acid, benzenepropanoic acid, 4-cyano, 4-cyanohydrocinnamic acid, benzenepropanoic acid,4-cyano, 3-4-cyanophenyl propionic acid, 3-p-cyanophenyl-propionic acid, 3-4-cyano-phenyl propionic acid
PubChem CID 9361742
IUPAC Name 3-(4-cyanophenyl)propanoic acid
SMILES C1=CC(=CC=C1CCC(=O)O)C#N

Specifications

Quantity 5 g
UN Number UN3439
Formula Weight 175.19
Percent Purity 96%
Chemical Name or Material 3-(4-Cyanophenyl)propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.