missing translation for 'onlineSavingsMsg'
Learn More

3-(4-chlorophenyl)-1H-pyrazol-5-amine, 97%, Thermo Scientific™

Product Code. 10354712
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10354712 10 g Amber Glass Bottle 10g
10551872 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10354712 Supplier Thermo Scientific Maybridge Supplier No. KM01548EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 78583-81-0
Molecular Formula C9H8ClN3
Molecular Weight (g/mol) 193.63
MDL Number MFCD00053046
InChI Key XQPBZIITFQHIDI-UHFFFAOYSA-N
Synonym 3-4-chlorophenyl-1h-pyrazol-5-amine, 5-4-chlorophenyl-1h-pyrazol-3-amine, 5-amino-3-4-chlorophenyl pyrazole, 5-4-chlorophenyl-2h-pyrazol-3-amine, 3-amino-5-4-chlorophenyl-1h-pyrazole, 5-amino-3-4-chlorophenyl-1h-pyrazole, 3-amino-5-4-chlorophenyl pyrazole, 5-4-chlorophenyl-2h-pyrazol-3-ylamine, 3-4-chlorophenyl pyrazole-5-ylamine, pubchem12126
PubChem CID 2735305
IUPAC Name 5-(4-chlorophenyl)-1H-pyrazol-3-amine
SMILES NC1=NNC(=C1)C1=CC=C(Cl)C=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 193.64
Percent Purity 97%
Chemical Name or Material 3-(4-Chlorophenyl)-1H-pyrazol-5-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.