missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Chlorophenoxy)pentane-2,4-dione, Tech., Thermo Scientific™

Product Code. 10263472
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10263472 10 g 10g
10427062 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10263472 Supplier Thermo Scientific Maybridge Supplier No. SPB03987EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 31168-10-2
Molecular Formula C11H11ClO3
Molecular Weight (g/mol) 226.656
MDL Number MFCD00204239
InChI Key QOOLFYCEZZCVSE-UHFFFAOYSA-N
Synonym 3-4-chlorophenoxy pentane-2,4-dione, 2,4-pentanedione,3-4-chlorophenoxy, 3-4-chlorophenoxy-2,4-pentanedione
PubChem CID 2799447
IUPAC Name 3-(4-chlorophenoxy)pentane-2,4-dione
SMILES CC(=O)C(C(=O)C)OC1=CC=C(C=C1)Cl

Specifications

Quantity 10 g
Linear Formula ClC6H4OCHC(O)CH32
Formula Weight 226.66
Percent Purity Tech.
Grade Technical
Chemical Name or Material 3-(4-Chlorophenoxy)pentane-2, 4-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.