missing translation for 'onlineSavingsMsg'
Learn More

3-[4-(Chloromethyl)phenyl]-1-methyl-1H-pyrazole, 97%, Thermo Scientific™

Product Code. p-3541015
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11350587

Brand: Thermo Scientific Maybridge CC23824CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 916766-83-1
Molecular Formula C11H11ClN2
Molecular Weight (g/mol) 206.673
MDL Number MFCD09702351
InChI Key YLJMPTWMRVXTQQ-UHFFFAOYSA-N
Synonym 3-4-chloromethyl phenyl-1-methyl-1h-pyrazole, 3-4-chloromethyl phenyl-1-methylpyrazole
PubChem CID 24229472
IUPAC Name 3-[4-(chloromethyl)phenyl]-1-methylpyrazole
SMILES CN1C=CC(=N1)C2=CC=C(C=C2)CCl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 206.67
Percent Purity 97%
Chemical Name or Material 3-[4-(Chloromethyl)phenyl]-1-methyl-1H-pyrazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.