missing translation for 'onlineSavingsMsg'
Learn More

3-[4-(Chloromethyl)phenoxy]-N,N-dimethylpropylamine hydrochloride, 90%, Thermo Scientific™

Product Code. 10481134
Click to view available options
Amber Glass Bottle; 1g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 769123-68-4
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Molecular Formula C12H19Cl2NO
MDL Number MFCD09702396
Quantity 1 g
Synonym 3-4-chloromethyl phenoxy-n,n-dimethylpropylamine hydrochloride, 3-4-chloromethyl phenoxy propyl dimethylamine hydrochloride, 3-4-chloromethyl phenoxy propyl dimethylamine, chloride, 3-4-chloromethyl phenoxy-n,n-dimethylpropan-1-amine hydrochloride, 3-4-chloromethyl phenoxy-n,n-dimethylpropan-1-amine-hydrogen chloride 1/1
InChI Key QJUMEHQGZPKXLM-UHFFFAOYSA-N
SMILES CN(C)CCCOC1=CC=C(C=C1)CCl.Cl
IUPAC Name 3-[4-(chloromethyl)phenoxy]-N,N-dimethylpropan-1-amine;hydrochloride
Molecular Weight (g/mol) 264.19
PubChem CID 24229611
Formula Weight 264.19
Percent Purity 90%
Chemical Name or Material 3-[4-(Chloromethyl)phenoxy]-N, N-dimethylpropylamine hydrochloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'